Imprimer
GL adapters for barrels, BOLA
Fournisseur: Bohlender
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
D737-24 D737-12 D736-24 D736-12
BOHLD737-24EA 69.4 EUR
BOHLD737-24 BOHLD737-12 BOHLD736-24 BOHLD736-12
GL adapters for barrels, BOLA
Barils, tonneaux, fûts
Adaptors made of PTFE/PP, for transition from female barrel thread R2" or Tri-Sure 2" to a GL 45 thread.
  • Universal chemical resistance
  • Temperature resistance from −200 to 250 °C
  • Autoclavable and can be used under vacuum