Imprimer
Pipette controller, Pipette Pump™, SP Bel-Art
Fournisseur: Bel-Art Products, a Part of SP
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
F378960000 F378970000 F378980000 F378990000
BELAF378960000EA 121 EUR
BELAF378960000 612-1922 612-1923 BELAF378990000
Pipette controller, Pipette Pump™, SP Bel-Art
Pipettes Pipeteurs
Pipette filler for glass and plastic pipettes up to 0,2, 2, 10 or 25 ml.
  • Pipettes fit smoothly into its flexible tapered chuck
  • Thumbwheel on side is rotated for precision aspirating or dispensing
  • Plunger can be pressed for quick emptying
  • Resistant to acids and alkalis
  • Autoclavable at 121 °C for 20 minutes (chuck should not be autoclaved)
These fillers provide safe, accurate and trouble-free pipetting with simple and one-handed operation.
Informations de commande: Replacement silicone rubber chuck and washers are available on request.