Imprimer
Tubing, PTFE, anti-static, explosion-proof, BOLA
Fournisseur: Bohlender
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
S1827-40 S1827-60 S1827-64 S1827-50 S1827-68 S1827-30 S1827-26 S1827-10 S1827-42 S1827-74
BOHLS1827-40EA 27.8 EUR
BOHLS1827-40 BOHLS1827-60 BOHLS1827-64 BOHLS1827-50 BOHLS1827-68 BOHLS1827-30 BOHLS1827-26 BOHLS1827-10 BOHLS1827-42 BOHLS1827-74
Tubing, PTFE, anti-static, explosion-proof, BOLA
Tuyaux Transfer Tubing
Electric conductive black tubing with antistatic compound made of pure PTFE and finest, highly pure carbon dust (less than 2,5%). Ideal for light-sensitive substances.
  • Extensive chemical resistance due to PTFE parts
  • Temperature resistance from −200 to +250 °C
  • Resistance of less than 1,000 kOhm
They have antistatic applications, used in explosive ambiance (explosion protection), also for transport of solvents or alcohols.