Imprimer
Stirrer shafts for magnetic stirrer heads, glass, BOLA
Fournisseur: Bohlender
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
C492-16 C492-24
BOHLC492-16EA 189 EUR
BOHLC492-16 BOHLC492-24
Stirrer shafts for magnetic stirrer heads, glass, BOLA
Stirrers Stirring Shafts
Stirrer shafts made of borosilicate glass with integrated counterbores for the three stud screws of the rotor of the magnetic stirrer head.
  • Resistant to chemicals
  • Temperature resistance from –200 to +250 °C
Ideal for use together with stirrer blades which can be fixed individually.