Imprimer
Reaction vessels, cylindrical, with flat flange
Fournisseur: KGW
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
TRGN7189-J15 TRGN7173 TRGN7174 TRGN7273 TRGN7198-20
KGWATRGN7189-J15EA 0 EUR
KGWATRGN7189-J15 KGWATRGN7173 KGWATRGN7174 KGWATRGN7273 KGWATRGN7198-20
Reaction vessels, cylindrical, with flat flange
Récipients de réaction Récipients cylindriques à réaction
Borosilicate glass.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With thermal jacket, DN15 flange and drain valve.