Imprimer
Accessoires pour distributeur de bouchons d'oreille à usage unique, uvex one 2 click
Fournisseur: UVEX
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
2112.003 2112.023 2112.118 2112.022
UVEX2112.003EA 56.4 EUR
UVEX2112.003 UVEX2112.023 UVEX2112.118 UVEX2112.022
Accessoires pour distributeur de bouchons d'oreille à usage unique, uvex one 2 click
Bouchons d'oreille Bouchons d'oreilles à usage unique
Refill packs and boxes for the uvex x-fit earplug dispenser "one 2 click".
uvex x-fit: The ergonomically shaped uvex x-fit disposable earplug is characterised by very high sound absorption levels, making it suitable for use in very loud environments. The uvex x-fit foam earplug has a sealed surface, preventing foreign bodies from penetrating. The soft foam offers real comfort, even when worn for extended periods. Ideal for use in very loud environments. SNR = 37 dB.
uvex com4-fit: With its compact, ergonomically shaped design, the uvex com4-fit is comfortable even for wearers withEn savoir plus