Imprimer
Reaction vessels, cylindrical, with thermal jacket, GL-18 olive and drain valve
Fournisseur: KGW
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
TRGN7229 TRGN7215 TRGN7207 TRGN7107
KGWATRGN7229EA 0 EUR
KGWATRGN7229 KGWATRGN7215 KGWATRGN7207 KGWATRGN7107
Reaction vessels, cylindrical, with thermal jacket, GL-18 olive and drain valve
Récipients de réaction Récipients cylindriques à réaction
Borosilicate glass.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With thermal jacket, GL-18 Olive tubing connection and drain valve.