Imprimer
Bottles, wide neck, square, with closure, SP Bel-Art
Fournisseur: Bel-Art Products, a Part of SP
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
F109041000 F109040500
BELAF109041000EA 277 EUR
BELAF109041000 BELAF109040500
Bottles, wide neck, square, with closure, SP Bel-Art
Flacons Flacons en plastique
Molded of high density polyethylene (HDPE).
  • Deep shelled, blue polypropylene closures
  • Ideal for storing or shipping samples
  • Good chemical and temperature resistance
Leakproof, chemical resistant bottles have heavy-threaded necks and tamper-proof, tear-lock closures.