Imprimer
Infrared heaters, IRB series
Fournisseur: Edmund Bühler GmbH
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
6063000 6060000 9066000
451-1502EA 744 EUR
451-1502 451-1500 451-1505
Infrared heaters, IRB series
Appareils chauffants Plaques chauffantes
The IRB infrared heaters heated up samples to 700 or 900 °C within a short time.
  • Ideal for use where unshielded flames are not allowed for safety reasons