Imprimer
COD test kits and consummables for AQUAfast COD colorimeters
Marques : Thermo Orion
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
CODHP0 CODH00 CODL00
662-1543EA 75.4 EUR
662-1543 705-0757 705-0758
COD test kits and consummables for AQUAfast COD colorimeters
Kits de tests environnementaux
Orion™ AQUAfast™ COD reagents in digestion tubes are designed for use with Orion™ AQUAfast™ colorimeters and Orion™ AquaMate Vis and UV-Vis spectrophotometers. Just add the sample to the COD digestion tube, digest sample in thermoreactor and measure.
  • Reagents are fully prepared and offer convenience and added safety for user
  • Reagents comply with EPA guidelnes
Digestion tubes contain the precise amount of reagent, are made of special optical glass, and fit the Orion™ AQUAfast™ COD165 thermoreactor.