Imprimer
Dimroth condensers for Soxleth extractors
Fournisseur: Lenz Laborglas GmbH & CO.KG
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
05246004 05246006 05246007 05246008 05246005
533-2121EA 134 EUR
533-2121 533-2123 533-2127 537-0032 533-2122
Dimroth condensers for Soxleth extractors
Condensers
DURAN® borosilicate glass.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Dimroth condensers for extractors, for condensing vapours. Water connection with two glass olives.