Imprimer
Dispositif de révélation par immersion pour les plaques CCM
Fournisseur: CAMAG
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
022.6606 022.6619 022.6622 022.6627 022.6628
552-0034EA 4880 EUR
552-0034 552-0035 552-0036 552-0037 552-0038
Dispositif de révélation par immersion pour les plaques CCM
Matériel de chromatographie Chromatographie, Pièces de rechange
This chromatogram immersion device is suitable for TLC and HPTLC plates up to 200×200 mm.
  • Uniform vertical speed, freely selectable between 30 mm/s and 50 mm/s
  • Immersion time selectable between 1 and 8 seconds and indefinitely
  • The device can be set to accommodate 100 mm and 200 mm plate height
  • Battery operated, independent of power supply
For proper execution of the dipping technique, the chromatogram must be immersed and withdrawn at a controlled uniform speed. By maintaining a well defined vertical speed and immersion time, derivatisation conditions can be standardised and 'tide marks', which can interfere with densitometric evaluation, are avoided.
Informations de commande: Supplied without dip tanks.