Imprimer
Distributeurs, close-it
Fournisseur: BURKLE
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
5303-9000 5303-9100
312-1156EA 163 EUR
312-1156 312-0083
Distributeurs, close-it
Distributeurs d'étiquettes et de rubans adhésifs
Dispensers for control seals labels.
  • Powerful strength of the adhesive
  • Two sizes are available
  • Reliably prevents products from leaking out after sampling
Particularly suited for the food and pharma industries.
Certifications: FDA guidelines.
Informations de commande: Two different sizes available.