Imprimer
Exhaust filters for barrels
Fournisseur: S.C.A.T.
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
108986 108987 108985
SCTV108986EA 499 EUR
SCTV108986 SCTV108987 SCTV108985
Exhaust filters for barrels
Filters Venting Filters
SCAT exhaust air filters for drums / barrels offer optimal protection against vapors and gases that can escape through opened drums. The high-quality activated carbon prevents clumping when in contact with aerosols and thus prevents the escape of toxic impurities and reliably binds volatile substances.
  • Reliable protection of breathing air in closed rooms and thus the health of employees
  • Guarantee highest quality and safety standards for laboratories and the handling of highly volatile chemicals
  • Contain a high-quality activated carbon granulate, which has the highest and most lasting absorption effect within the guaranteed lifetime
The SCAT exhaust air filters are available in sizes XL (recommended for containers up to 100 L and a service life of six months) and XXL (recommended for containers of 100 L and above and a service life of 12 months). An indicator always shows the remaining time until the next filter change. By using adapters all thread sizes can be easily connected.
Certifications: Conform to the requirements of flammability classification V-0 (UL-94).
Cdt: If the filters are packed and stored in a dry place, there is then no known exact expiry date. After several years, the carbon will gradually coagulate and the overall surface area will change, but the filter can still be used.
Attention: Exchange your SCAT exhaust filters V2.0 regularly and within the given lifetime. Recommended service life when using LC-typical organic solvents and solvent mixtures.