Imprimer
UV rechargeable lamps, 26 P series
Fournisseur: UVP ULTRA VIOLET PRODUCTS
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
95-0181-02 95-0186-02 95-0187-02
UVPA95-0181-02EA 834 EUR
UVPA95-0181-02 UVPA95-0186-02 UVPA95-0187-02
UV rechargeable lamps, 26 P series
Éclairages Lampes UV
Rechargeable 6-watt 26P series lamps are available in three dual-tube models. The lamps are equipped with an internal gel based, lead-acid battery, which can be charged for field operation from a standard wall outlet. Model UVSL-26P can operate with both long and shortwave on at the same time.
Optional 12 V adapter operates the lamp from a car cigarette lighter.
W×D×H: 76×140×249 mm
Weight: 1,8 kg