Imprimer
Barboteurs avec tête, VitraPOR®
Fournisseur: ROBU GLASFILTER
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
40252 40254 40101 40100 40102 40103 40104 40250 40251 40253
ROBU40252EA 116 EUR
ROBU40252 ROBU40254 ROBU40101 ROBU40100 ROBU40102 ROBU40103 ROBU40104 ROBU40250 ROBU40251 ROBU40253
Barboteurs avec tête, VitraPOR®
Flacons Flacons-laveurs
Borosilicate glass.
  • With sintered glass disc, porosity grade 0/1/2/3/4
  • All system components can be ordered separately
  • Excellent chemical and temperature resistance
Consists of the gas washing bottle with NS 29/32 standard ground joint socket and the insert with NS 29/32 cone joint and two glass tubing connectors with 11 mm external diameter.