Imprimer
Distributeurs de lingettes, zetMatic®
Fournisseur: Zellstoff-Vertriebs-GmbH & Co. KG
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
60955 60953
115-2592EA 254 EUR
115-2592 115-2593
Distributeurs de lingettes, zetMatic®
Distributeurs de lingettes
Floor-standing dispensers made of steel with an aluminum powder coating. Suitable for rolls with a width of max. 34 or 42 cm.
  • Large model also features a waste bag holder and wheels for flexible usage
  • Wipe rolls can be easily exchanged