Imprimer
Dropping bottles, glass, Wheaton®
Fournisseur: DWK Life Sciences
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
W211735 W211734 W211754 W211755
WHEAW211735EA 166 EUR
WHEAW211735 WHEAW211734 WHEAW211754 WHEAW211755
Dropping bottles, glass, Wheaton®
Flacons Flacons compte-gouttes
Manufactured from soda-lime glass material.
  • Available in clear or amber glass format
  • Amber coloured to protect light sensitive substanzes
  • Not autoclavable
Dropping bottles delivers stains or indicator solutions drop by drop. Complete with a glass stopper, a partial turn of the ground glass stopper seals the bottle.
Certifications: Material conforms to USP Type III requirements.