Imprimer
Culture vials with cap, WHEATON®
Fournisseur: DWK Life Sciences
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
225142 225144 225145
218-2252EA 325 EUR
218-2252 218-2250 218-2251
Culture vials with cap, WHEATON®
Fioles
These culture vials feature deep skirted screw caps to allow safer, more dependable handling when working with infectious materials. They are manufactured from 33 low extractable borosilicate glass that conforms to USP Type I and ASTM E 438 Type I, Class A requirements.
  • Pre-attached deep skirted solid-top black phenolic screw cap with 14B rubber liner
Cdt: Vials are packaged in corrugated trays with partitions.