Imprimer
Arsine generator apparatus, modified, WHEATON®
Fournisseur: DWK Life Sciences
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
180023 180024
WHEA180023EA 212 EUR
WHEA180023 WHEA180024
Arsine generator apparatus, modified, WHEATON®
Digestion Systems Trace Element Detection Apparatus
Modified version for arsine analysis which eliminates the cumbersome ball and socket joint connecting the scrubber and absorber tubes which conforms to ASTM / USP and EPA specifications.
The freestanding system has a standard taper 24/40 Clear-Seal™ joint. Manufactured from 33 low extractable borosilicate glass that conforms to USP Type I and ASTM E 438 Type I, Class A requirements.