Imprimer
Filtres
Fournisseur: OLYMPUS OPTICAL
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
N1977500 N1977600 N1977800 N1977900 N1978000 N1978100 N1978200 N1978300 N1978400
630-0946EA 0 EUR
630-0946 630-0947 630-0948 630-0949 630-0957 630-0958 630-0959 630-0960 630-0961
Filtres
Filtres Filtres optiques
Filter units, ranging from UV excitation to RFP, respond to applications using various fluorescent dyes and protein.
High quality (HQ) filters have an edge steepness and high transmission that efficiently detect the light of fluorescence to enhance and capture brighter fluorescence images in precise detail.