Imprimer
Calibration weight sets, OIML class F1
Fournisseur: Sartorius Balances
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
YCS31-653-00 YCS31-613-02 YCS31-613-00 YCS31-653-02 YCS01-653-00 YCS01-653-02 YCS31-713-00 YCS31-713-02 YCS01-523-02 YCS01-613-00 YCS01-613-02 YCS01-513-00 YCS01-513-02 YCS01-253-AC02 YCS01-253-AC00 YCS31-513-AC00 YCS31-513-AC02 YCS31-653-AC02 YCS31-653-AC00 YCS31-613-AC00
SARBYCS31-653-00EA 4140 EUR
SARBYCS31-653-00 SARBYCS31-613-02 SARBYCS31-613-00 SARBYCS31-653-02 SARBYCS01-653-00 SARBYCS01-653-02 SARBYCS31-713-00 SARBYCS31-713-02 SARBYCS01-523-02 SARBYCS01-613-00 SARBYCS01-613-02 SARBYCS01-513-00 SARBYCS01-513-02 SARBYCS01-253-AC02 SARBYCS01-253-AC00 SARBYCS31-513-AC00 SARBYCS31-513-AC02 SARBYCS31-653-AC02 SARBYCS31-653-AC00 SARBYCS31-613-AC00
Calibration weight sets, OIML class F1
Poids Poids d'étalonnage
Class F1 weight sets include aluminium (1 to 5 mg) or nickel silver (10 to 500 mg) or stainless steel (1 g and higher) weights.