Imprimer
Calibration weight sets, OIML class E1
Fournisseur: Sartorius Balances
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
YCS31-651-00 YCS31-651-02 YCS31-711-00 YCS31-611-00 YCS31-611-02 YCS31-711-02 YCS011-521-02 YCS011-351-02 YCS011-511-00 YCS011-511-02 YCS011-521-00 YCS011-611-00 YCS011-611-02 YCS011-351-00 YCS011-651-00 YCS011-651-02
SARBYCS31-651-00EA 11240 EUR
SARBYCS31-651-00 SARBYCS31-651-02 SARBYCS31-711-00 SARBYCS31-611-00 SARBYCS31-611-02 SARBYCS31-711-02 SARBYCS011-521-02 SARBYCS011-351-02 SARBYCS011-511-00 SARBYCS011-511-02 SARBYCS011-521-00 SARBYCS011-611-00 SARBYCS011-611-02 SARBYCS011-351-00 SARBYCS011-651-00 SARBYCS011-651-02
Calibration weight sets, OIML class E1
Poids Poids d'étalonnage
These E1 class weight sets from 1 mg to 10 kg are made of non-magnetisable, stainless steel.
  • Density of 8,0 g/dm³
  • Knob weights highly polished
  • Highly corrosion-resistant
Informations de livraison: Up to 20 g supplied in a plastic case; from 50 g and up in a wooden case; from 1 kg and up, additionally supplied with gloves.