Imprimer
Cordless pumps, BOLA
Fournisseur: Bohlender
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
G870-01 G870-11
223-0039EA 725 EUR
223-0039 223-0040
Cordless pumps, BOLA
Pompe Pompes pour conteneur/transfert
Cordless pumps are made of PP, PTFE and Hastelloy®. They are applicable for pumping low viscous liquids.
  • Powerful pumping capacity of up to six litres per minute free flowing
  • Compact construction and easy handling
  • Good chemical resistance
  • Very light weight (500 g including batteries)
  • Low-risk pumping
Temperature resistance from 5 to 60 °C.
They are also suitable for narrow mouth vessels with ground joint 29/32 or thread GL 45, carboys or barrels.
Informations de livraison: With two batteries 1,5 V