Imprimer
RNase A
Marques : OMEGA BIO-TEK
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
AC118 AC117
AC118EA 184 EUR
AC118 AC117
RNase A
Enzymes
Ribonuclease A (RNase A) is endonuclease-free and quality controlled for use in plasmid purification procedures for digestion of RNA.