Imprimer
Sub-surface samplers, Wheaton® Grab Sampler® II
Fournisseur: DWK Life Sciences
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
990350 990400 990450
WHEA990350EA 1100 EUR
WHEA990350 WHEA990400 WHEA990450
Sub-surface samplers, Wheaton® Grab Sampler® II
Échantillonneurs Préleveurs de liquides
The high density PP head holds a narrow mouth borosilicate jar.
  • Remote valve that can be opened by pulling and releasing the finger ring on the handle
  • Aluminum sections can be broken down for easy transport
  • Three lengths available
Certifications: EPA requirements for water and waste water sampling.