Imprimer
Stirrer shafts, with four bladed propeller, BOLA
Fournisseur: Bohlender
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
BOLA
BOHLC484-36EA 771 EUR
BOHLC484-36 BOHLC484-34 BOHLC484-18 BOHLC484-40 BOHLC484-50 BOHLC484-44 BOHLC484-22 BOHLC484-32
Stirrer shafts, with four bladed propeller, BOLA
Agitateurs Axes d'agitation
Stirrer shafts made of PTFE-jacketed stainless steel and the propeller completely made of PTFE.
  • With 45° angled angular blades
  • Chemical resistant
  • Temperature resistance from –200 to +250 °C