Imprimer
Stirrer shafts, with mini-propeller, BOLA
Fournisseur: Bohlender
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
C482-24 C482-12
BOHLC482-24EA 210 EUR
BOHLC482-24 BOHLC482-12
Stirrer shafts, with mini-propeller, BOLA
Agitateurs Axes d'agitation
Stirrer shafts made of PTFE-jacketed stainless steel and the propeller completely made of PTFE.
  • Ideal for stirring in narrow mouth vessels or in vessels with ground joint openings
  • With three 45° angled angular blades
  • Chemical resistant
  • Temperature resistance from –200 to +250 °C