Imprimer
Screw caps for high recovery vials, E-Z Ex-Traction™, WHEATON®
Fournisseur: DWK Life Sciences
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
W242710 W240832 W240841 W240831 W240830 W240846 W240840 W240844 W240836 W240850 W240843 W240848 W240851 W240842
WHEAW242710EA 205 EUR
WHEAW242710 WHEAW240832 WHEAW240841 WHEAW240831 WHEAW240830 WHEAW240846 WHEAW240840 WHEAW240844 WHEAW240836 WHEAW240850 WHEAW240843 WHEAW240848 WHEAW240851 WHEAW240842
Screw caps for high recovery vials, E-Z Ex-Traction™, WHEATON®
Capuchons Bouchons pour tubes à essais
Choice of black phenolic, white polypropylene or urea screw caps
  • PTFE faced (14B) styrene-butadiene rubber liner provides a totally inert inner seal and surface facing the sample or product
  • Bonded PTFE faced silicone liner ideal for storage of volatile solvents, sensitive compounds and corrosive chemicals