Imprimer
Blocks for RapidVap® Vertex™ dry block evaporators
Fournisseur: LABCONCO
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
7321200 7321300 7321600 7322000 7322800 7322801 7324100
LBCP7321200EA 2790 EUR
LBCP7321200 LBCP7321300 LBCP7321600 LBCP7322000 LBCP7322800 LBCP7322801 LBCP7324100
Blocks for RapidVap® Vertex™ dry block evaporators
Chauffe-blocs Blocs pour bains
All RapidVap® Vertex™ Dry Block evaporators require a Block (not included). Blocks of solid aluminum include stainless steel handles to lift the block in and out of the evaporator. A 0,21" (0,5 cm) diameter port in the upper left hand corner of the block is provided for insertion of the temperature probe to monitor upper block temperature.
Informations de commande: Select a block that closely matches the outside diameter of your sample container. Glassware is required (not included).