Imprimer
Büchner funnels, single piece, SP Bel-Art
Fournisseur: Bel-Art Products, a Part of SP
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
F146110000 F146080000 F146090000
BELAF146110000EA 119 EUR
BELAF146110000 BELAF146080000 BELAF146090000
Büchner funnels, single piece, SP Bel-Art
Entonnoirs Entonnoirs pour la filtration Entonnoirs Büchner
HDPE, with Fritware® porous plates.
  • Can be used with or without filter paper
  • For use below 52 °C
  • To be used with Vac-Ring™ filter seal for quick filter set up
These fast flowing funnels with sealed-in PE filter plate of coarse porosity (90 to 130 µm) offer good retention and fast flow under vacuum.