Imprimer
Test sieves, ASTM E 11, 200×25 mm
Fournisseur: Retsch GmbH
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
60.123.000020 60.123.000025 60.123.000032 60.123.000038 60.123.000045 60.123.000053 60.123.000063 60.123.000075 60.123.000090 60.123.000106 60.123.000125 60.123.000150 60.123.000180 60.123.000212 60.123.000250 60.123.000300 60.123.000355 60.123.000425 60.123.000500 60.123.000600 60.123.000710 60.123.000850 60.123.001000 60.123.001180 60.123.001400 60.123.001700 60.123.002000 60.123.002360 60.123.002800 60.123.003350 60.123.004000 60.123.004750 60.123.005600 60.123.006300 60.123.006700 60.123.008000 60.123.009500 60.123.011200 60.123.012500 60.123.013200 60.123.016000 60.123.019000 60.123.022400 60.123.025000 60.123.026500 60.123.031500 60.123.037500 60.123.045000 60.123.050000 60.123.053000 60.123.063000 60.123.075000 60.123.090000 60.123.100000 60.123.106000 60.123.125000
510-5257EA 1070 EUR
510-5257 510-5258 510-5259 510-5260 510-5261 510-5262 510-5263 510-5264 510-5265 510-5266 510-5267 510-5268 510-5269 510-5270 510-5271 510-5272 510-5273 510-5274 510-5275 510-5276 510-5277 510-5278 510-5279 510-5280 510-5281 510-5282 510-5283 510-5284 510-5285 510-5286 510-5287 510-5288 510-5289 510-5290 510-5291 510-5292 510-5293 510-5294 510-5295 510-5296 510-5297 510-5298 510-5299 510-5300 510-5301 510-5302 510-5303 510-5304 510-5305 510-5306 510-5307 510-5308 510-5309 510-5310 510-5311 510-5312
Test sieves, ASTM E 11, 200×25 mm
Tamis Tamis analytiques
Sieves made from 304 stainless steel are individually laser engraved with clear and precise labelling for full traceability. They are fully compatible with Retsch's existing products and can be combined with other sieves without problems.
  • Sieve bodies are made in one piece and the absence of hollow spaces prevents cross-contamination
  • High corrosion resistance and easy to clean
  • Maximum stability and optimal sealing of the sieve stack, 15% less weight than traditional sieves
Sieve meshes, frames and labeling comply with standards. The sieves are tested 5 times and are delivered with test report.
Certifications: An individual inspection certificate for test materials monitoring according to ASTM E 11 is also available on request.
Informations de commande: Sieves and accessories of other dimensions are available on request. Also available with perforated plate, round or square. For a full overview, please go to the website.