Imprimer
PCR Mycoplasma Test Kit II
Marques : PanReac AppliChem
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
A8994.0050 A8994.0025 A8994.0100
A8994.0050EA 998 EUR
A8994.0050 A8994.0025 A8994.0100
PCR Mycoplasma Test Kit II
Microbiology Test Systems Microorganism Tests
Lyophilised PCR mix for the detection of mycoplasma in cell culture by conventional PCR.
Meets criteria of section 2.6.7 of Ph. Eur.
Shipped at room temperature.
Informations de livraison: Supplied without Taq DNA polymerase. Kit contains PCR primer nucleotide mix, reaction buffer solution, water (PCR grade), positive template control, and internal control DNA.