Imprimer
Beakers, graduated, tall form, PP, SP Bel-Art
Fournisseur: Bel-Art Products, a Part of SP
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
F289910000 F289920000 F289900000 F289940000 F289930000 F289950000
BELAF289910000EA 39 EUR
BELAF289910000 BELAF289920000 BELAF289900000 BELAF289940000 BELAF289930000 BELAF289950000
Beakers, graduated, tall form, PP, SP Bel-Art
Béchers
Made of PP, translucent.
  • Use with wet or dry ingredients
  • Chemical resistant and shatterproof
  • Metric gradations molded on outside wall
  • Translucent polypropylene construction
  • Heat resistant to 121ºC (250ºF); steam autoclavable
Available in a large selection of sizes, these pitchers are ideal for general laboratory use and offer well designed spouts and large open handles for easy stacking.