Imprimer
Bottles, wide neck, square, WHEATON®
Fournisseur: DWK Life Sciences
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
W216871 W216882 W216879 W216880 W216881 W216873 W216874 W216875 W216876 W216877 W216878 W216872 W216889 W216898
WHEAW216871EA 75.9 EUR
WHEAW216871 215-1680 215-1697 215-1698 215-1699 WHEAW216873 WHEAW216874 WHEAW216875 WHEAW216876 WHEAW216877 WHEAW216878 WHEAW216872 WHEAW216889 WHEAW216898
Bottles, wide neck, square, WHEATON®
Flacons Flacons en verre
Square shape glass bottles provides maximum storage space.
  • Ideal for solvent, chemical or sample storage
  • Clear glass helps for easy sample observation
  • Made of type III soda-lime glass