Imprimer
Bottles, wide neck, with screw cap, WHEATON®
Fournisseur: DWK Life Sciences
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
W216924 W216934 W216935 W216937 W216925 W216926 W216933 W216929 W216931 W216930 W216936
215-4428EA 41.1 EUR
215-4428 WHEAW216934 WHEAW216935 WHEAW216937 215-4429 215-4430 215-4051 215-4047 215-4049 215-4048 WHEAW216936
Bottles, wide neck, with screw cap, WHEATON®
Flacons Flacons en verre
Made of type III soda-lime glass, clear.
  • Largest bottles available
  • Very good chemical resistance
  • PP screw cap with poly-vinyl liner or PTFE lined screw cap
Ideal for general, small to large volume storage. Hazardous communication label included with each case of bottles allows for content ID and safety notification. Tamper-evident seal included with each case of bottles keeps bottle sealed which protecting contents from accidental misuse.