Imprimer
Ground joint tube fittings, BOLA
Fournisseur: Bohlender
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
B304-24 B304-22 B304-20 B304-16 B304-10
BOHLB304-24EA 106 EUR
BOHLB304-24 BOHLB304-22 BOHLB304-20 BOHLB304-16 BOHLB304-10
Ground joint tube fittings, BOLA
Adaptateurs Raccords rodés
PTFE, white.
  • Temperature resistance: −200 to +250 °C
  • Universal chemical resistance
  • Autoclavable
For connecting tubes, hard-walled tubing, thermometers. The sealing rings on the outside of the cone prevent sticking of the ground joints and improve the sealing. Integrated special nut (Safe-Lab) for easy locking and unlocking of the ground joint.