Imprimer
VWR®, Feuilles adhésives pour PCR
Fournisseur: VWR Collection
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
391-1256EA 173 EUR
391-1256 391-1257
VWR®, Feuilles adhésives pour PCR
Scellés Microplaque Sceaux
These soft, non permeable 63,5 µm thick aluminium foils, with strong medical grade adhesive, eliminate the need for heat sealing devices or mats during thermal cycling. Compared to other aluminium foils, these foils have less tendency to roll back on themselves when removing the backing paper, and fit well to the plate during application.
  • Recommended for temperatures from −80 to +120 °C
  • Easily pierceable with pipette tips and robotic probes
  • Excellent vapour barrier, virtually no sample evaporation
  • Certified free from DNase, RNase and nuclease
Each foil L×D: 142,9×82,6 mm with sufficient sealing area for all PCR plates.
Longueur sans les pattes d’extrémité : 125,4 mm.
Cdt: Sterile product is packed in tamper-evident bags of 25/bag.