Imprimer
Slide holders, DrainRack™
Fournisseur: Simport Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
M710-100B M710-100G M710-100P M710-100W M710-100Y
631-1910EA 47.7 EUR
631-1910 631-1911 631-1912 631-1913 631-1914
Slide holders, DrainRack™
Slide Holders
This rugged tray used as a drain rack can hold up to 200 microscope slides in 100 individually numbered slots. All slides are stored upright for easier insertion and removal.
To save space, the number of slides held can be doubled simply by placing two slides in each slot, making room for 200 slides instead of 100.
Barcodes can be read without having to remove the slides from the box.