Imprimer
Columns for Organic Acid Analysis
Fournisseur: TRANSGENOMIC
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
ICE-99-2359 ICE-99-2369 ICE-99-9753
TRNGICE-99-2359EA 0 EUR
TRNGICE-99-2359 TRNGICE-99-2369 TRNGICE-99-9753
Columns for Organic Acid Analysis
Colonnes pour chromatographie
Ion exclusion is the preferred method for the separation of weakly ionisable species such as organic acids and alcohols.