Imprimer
Magnetic Stirrers, Spherical
Fournisseur: COWIE
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
442-9801EA 21.8 EUR
442-9801
Magnetic Stirrers, Spherical
PTFE, specifically designed for use in tubes and eccentric stirring.
  • Good chemical resistance
  • Temperature resistant, suitable for use at high and low temperatures (-200 to +250 °C)