Imprimer
Absorbent for mercury, Chemizorb® Hg
Fournisseur: Merck
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
1.01569.0001 1.12576.0001
1.01569.0001EA 169.5 EUR
1.01569.0001 1.12576.0001
Absorbent for mercury, Chemizorb® Hg
Spill Control Materials
Chemizorb® Hg is an all-inclusive set of reagents and auxiliaries for the safe and complete removal of drops of mercury and of traces of elemental mercury.
  • Sufficient for the safe and simple decontamination of an area of roughly one square metre
  • Chemizorb Hg does not affect skin, gloves should be worn to avoid contact with mercury
  • Mercury containing waste material must be disposed of as special waste in accordance with the official regulations
Informations de livraison: Pack contains 500 g reagent 1 and 100 ml reagent 2, plus accessories in plastic case.
Attention: Merck products are not available from Avantor in all countries. Please check with your local distributor.