Imprimer
Pathology knife, Black Diamond™
Fournisseur: CELLPATH
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
CAA-0401-01A CAA-0601-01A CAA-0801-01A CAA-1001-01A CAA-9901-01A
720-0010EA 101 EUR
720-0010 720-0011 720-0012 720-0013 720-0015
Pathology knife, Black Diamond™
Couteaux
Disposable cartilage knife with Sheffield stainless steel double honed blade and molded handle with finger guard.
  • Excellent alternative to resharpening autopsy knives
  • Complete disposal of knife and handle eliminates problems of replacement blade fit and ensures contamination-free environment
  • Available in five different types and lengths (cartilage, trimming, macro, lung and brain knives) to enable pathologist to easily select optimum size of knife for any purpose
Informations de livraison: Supplied in packs of 10 or an assorted pack containing two knives of each type and length