Imprimer
Identification cards, multi-coloured, 96-well and 384-well
Fournisseur: DIVERSIFIED BIOTECH
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
WELL-384H WELL-384V WELL-96H WELL-96V
613-1453EA 182 EUR
613-1453 613-1454 DIVEWELL-96H DIVEWELL-96V
Identification cards, multi-coloured, 96-well and 384-well
Well Plate Stands and Well Orienters
Visual aid for pipetting and identification of samples in microplates. Fits most standard 96-well and 384-well plates.
  • Bright colours make it easy to distinguish between columns and rows
Can be used with the Pipette Guide for convenient viewing.