Imprimer
Conteneurs pour échantillons, avec couvercle
Fournisseur: Simport Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
C581-500 C580-3 C580-4 C580-5 C581-1L C581-300 C581-2600 C581-5700
216-1005EA 149 EUR
216-1005 216-1001 216-1002 216-1003 SIMPC581-1L SIMPC581-300 SIMPC581-2600 SIMPC581-5700
Conteneurs pour échantillons, avec couvercle
Récipients de collecte d'échantillon Conteneurs à usage universel
These containers and closures are made of PP. They are ideal for collection, transport and storage of fluids, powders, solids, pathology specimens and hazardous samples.
  • Disposable
  • Resist temperatures from −20 to + 80 °C
Tight-fitting lids prevent leaks and odors. Certain models listed below have a tight-fitting lid. Also offered with an incorporate tamperproof lid which is opened by first removing tab on side of lid, then by lifting the cover.
The 2600 and 5700 ml sizes are provided with a plastic handle to facilitate handling.
Attention: *Le(s) produit(s) sur cette page sont hors du champ d'application de la directive européenne 2019/904