Imprimer
VWR®, Funnels with Long Stem
Fournisseur: VWR Collection
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
221-0204EA 15.7 EUR
221-0204 221-0205 221-0206 221-0207 221-0208
VWR®, Funnels with Long Stem
Funnels Filtration Funnels Filter Funnels
PTFE, white.
  • Temperature resistance: –200 to +280 °C
  • Totally chemically inert
  • Super smooth non-stick internal finish
Conical opening with long stem.