Imprimer
Roller bottles, WHEATON®
Fournisseur: DWK Life Sciences
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
348273 348252 348253 348256 348258 348522 348528 348272 348524
734-3022EA 433 EUR
734-3022 734-3017 734-3018 734-3019 734-3020 734-3023 734-3025 734-3021 734-3024
Roller bottles, WHEATON®
Bottles Roller Bottles
Rollers bottles are specifically designed for adherent cell roller culture
  • Bottles withstands repeated wet or dry sterilization cycles and are reusuable
  • These culture vessels are made to exacting tolerances in order to provide higher cell yields
  • Made of USP Type I borosilicate glass
Attention: *The product(s) on this page are outside the scope of the EU Directive 2019/904