Imprimer
Tubing distributors, miniature, BOLA
Fournisseur: Bohlender
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
F710-07 F710-09 F710-05 F710-03 F710-01 F710-11
BOHLF710-07EA 93.4 EUR
BOHLF710-07 BOHLF710-09 BOHLF710-05 BOHLF710-03 BOHLF710-01 BOHLF710-11
Tubing distributors, miniature, BOLA
Raccords et connecteurs
Miniature distributors, made of PTFE, with female thread UNF ¼" 28 G, blockshaped, with holes for mounting (2ר 3,5 mm).
  • Temperature resistance: from −200 to +250 °C
  • Universal chemical resistance
  • Pressure: 30 bar