Imprimer
Sample spoons, Sterileware®, SP Bel-Art
Fournisseur: Bel-Art Products, a Part of SP
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
F367400001 F367400003 F367400005
231-0616EA 347 EUR
231-0616 231-0618 231-0621
Sample spoons, Sterileware®, SP Bel-Art
Cuillères
PS, white. These economical spoons are ideal for retrieving small, specific volume samples of powders or liquids. Use a separate spoon for each material sampled to avoid cross contamination.
  • Unique flat bottom shape allows them to be used as weighing boats for additional sampling accuracy
  • Made from white, FDA grade high-impact PS for durability and stiffness when sampling packed powders
  • Sterile spoons are individually sealed and sterilised in a polyethylene bag ready for one time use and disposal
  • Lot stamped for accurate lot tracing and shipped with a Certificate of Processing for Sterilization