Imprimer
Magnetic stirring bars, PTFE, elliptical, Spinbar®, SP Bel-Art
Fournisseur: Bel-Art Products, a Part of SP
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
F371300001 F371300112 F371300114 F371300038 F371300034
BELAF371300001EA 0 EUR
BELAF371300001 442-9521 BELAF371300114 BELAF371300038 BELAF371300034
Magnetic stirring bars, PTFE, elliptical, Spinbar®, SP Bel-Art
Agitateurs Barreaux magnétiques
These PTFE covered egg-shaped magnetic stirring bars have their own pivot point without a separate ring or abrupt change in contour. Designed especially for use in round-bottom vessels, they spin unrestricted by the contours of the vessel.